missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Difluoromethoxy)benzenesulfonamide 98.0+%, TCI America™
$100.67 - $400.78
Chemical Identifiers
| CAS | 874781-09-6 |
|---|---|
| Molecular Formula | C7H7F2NO3S |
| Molecular Weight (g/mol) | 223.19 |
| MDL Number | MFCD03407973 |
| InChI Key | BBBQHNZWCPUJNW-UHFFFAOYSA-N |
| PubChem CID | 24820749 |
| IUPAC Name | 4-(difluoromethoxy)benzene-1-sulfonamide |
| SMILES | NS(=O)(=O)C1=CC=C(OC(F)F)C=C1 |
Chemical Identifiers
| 874781-09-6 | |
| 223.19 | |
| BBBQHNZWCPUJNW-UHFFFAOYSA-N | |
| 4-(difluoromethoxy)benzene-1-sulfonamide |
| C7H7F2NO3S | |
| MFCD03407973 | |
| 24820749 | |
| NS(=O)(=O)C1=CC=C(OC(F)F)C=C1 |
Specifications
| 874781-09-6 | |
| White-Yellow | |
| MFCD03407973 | |
| BBBQHNZWCPUJNW-UHFFFAOYSA-N | |
| 4-(difluoromethoxy)benzene-1-sulfonamide | |
| 24820749 | |
| ≥98.0% (HPLC,N) | |
| 4-(Difluoromethoxy)benzenesulfonamide |
| 113°C | |
| C7H7F2NO3S | |
| 1 g | |
| NS(=O)(=O)C1=CC=C(OC(F)F)C=C1 | |
| 223.19 | |
| 223.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No