missing translation for 'onlineSavingsMsg'
Learn More

4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluorobiphenyl 98.0+%, TCI America™

Catalog No. d51295g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D51295G 5 g
D51291G 1 g
2 options

Catalog No. D51295G

Supplier: TCI America D51295G

Only null left

Chemical Identifiers

CAS 303186-19-8
Molecular Formula C21H13F7O
Molecular Weight (g/mol) 414.32
MDL Number MFCD22376741
InChI Key ZTUBOAYBUBUTPH-UHFFFAOYSA-N
PubChem CID 58402371
IUPAC Name 4-[difluoro(3,4,5-trifluorophenoxy)methyl]-4'-ethyl-3,5-difluoro-1,1'-biphenyl
SMILES CCC1=CC=C(C=C1)C1=CC(F)=C(C(F)=C1)C(F)(F)OC1=CC(F)=C(F)C(F)=C1

Specifications

Color White
Quantity 5 g
Formula Weight 414.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-4′-ethyl-3,5-difluorobiphenyl