missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminobenzoic Acid 98.0+%, TCI America™

Catalog No. D24935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24935G 5 g
D249325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24935G Supplier TCI America Supplier No. D24935G

Chemical Identifiers

CAS 5429-28-7
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00002538
InChI Key LNYTUARMNSFFBE-UHFFFAOYSA-N
Synonym 4-diethylamino benzoic acid, benzoic acid, 4-diethylamino, 4-n,n-diethylaminobenzoic acid, p-diethylamino benzoic acid, 4-diethylaminobenzoicacid, acmc-209lfa, 4-diethylamino benzoic, p-diethylaminobenzoic acid, 4-diethylamino benzoicacid
PubChem CID 79480
IUPAC Name 4-(diethylamino)benzoic acid
SMILES CCN(CC)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diethylaminobenzoic Acid