missing translation for 'onlineSavingsMsg'
Learn More

4'-Diethylaminoacetophenone 98.0+%, TCI America™

Catalog No. D20845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20845G 5 g
D20841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20845G Supplier TCI America Supplier No. D20845G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5520-66-1
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00043651
InChI Key HMIBQFXWSUBFTG-UHFFFAOYSA-N
Synonym 4'-diethylaminoacetophenone, p-diethylaminoacetophenone, 4-diethylaminoacetophenone, 1-4-diethylamino phenyl ethanone, ethanone, 1-4-diethylamino phenyl, 1-acetyl-4-diethylamino benzene, acmc-1aspz, 4-acetyl-n,n-diethylaniline
PubChem CID 138529
IUPAC Name 1-[4-(diethylamino)phenyl]ethan-1-one
SMILES CCN(CC)C1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 49°C
Quantity 5 g
Formula Weight 191.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Diethylaminoacetophenone
TSCA No
Recommended Storage Refrigerator