accessibility menu, dialog, popup

UserName

CH3NCH3CH3O

CAS RN 5520-66-1

IUPAC Name: 1-[4-(diethylamino)phenyl]ethan-1-one
Synonyms: 1-[4-(diethylamino)phenyl]ethanone
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: HMIBQFXWSUBFTG-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1)C(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results