missing translation for 'onlineSavingsMsg'
Learn More

4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran 95.0+%, TCI America™

Catalog No. D2849500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2849500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2849500MG Supplier TCI America Supplier No. D2849500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51325-91-8
Molecular Formula C19H17N3O
Molecular Weight (g/mol) 303.365
InChI Key YLYPIBBGWLKELC-RMKNXTFCSA-N
Synonym unii-9s1wxc1zgn, dcm dye, dcm1, 9s1wxc1zgn, 4-dicyanomethylene-2-methyl-6-4-dimethylaminostyryl-4h-pyran, 4-dicyanomethylene-2-methyl-6-4-dimethylamino styryl-4h-pyran, e-2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile, 2-2-4-dimethylamino phenyl ethenyl-6-methyl-4h-pyran-4-ylidene propanedinitrile, 2-2-4-dimethylamino styryl-6-methyl-4h-pyran-4-ylidene malononitrile
PubChem CID 688222
IUPAC Name 2-[2-[(E)-2-[4-(dimethylamino)phenyl]ethenyl]-6-methylpyran-4-ylidene]propanedinitrile
SMILES CC1=CC(=C(C#N)C#N)C=C(O1)C=CC2=CC=C(C=C2)N(C)C

Specifications

Melting Point 220°C
Quantity 500 mg
UN Number 1325
Formula Weight 303.37
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dicyanomethylene)-2-methyl-6-(4-dimethylaminostyryl)-4H-pyran
RTECSNumber OO3746100
TSCA Yes