missing translation for 'onlineSavingsMsg'
Learn More

4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran 98.0+%, TCI America™

Catalog No. p-7127504
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27911G 1 g
D27915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27911G Supplier TCI America Supplier No. D27911G
Only null left

Chemical Identifiers

CAS 28286-88-6
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD00181519
InChI Key XYBUCJYJVULPHW-UHFFFAOYSA-N
Synonym 2-(2,6-Dimethyl-4H-pyran-4-ylidene)propanedinitrile
PubChem CID 119915
IUPAC Name 2-(2,6-dimethyl-4H-pyran-4-ylidene)propanedinitrile
SMILES CC1=CC(C=C(C)O1)=C(C#N)C#N

Specifications

Melting Point 193°C
Quantity 1 g
UN Number 3439
Formula Weight 172.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dicyanomethylene)-2,6-dimethyl-4H-pyran