missing translation for 'onlineSavingsMsg'
Learn More

4-(Dibutylamino)salicylaldehyde 96.0+%, TCI America™

$393.32 - $1377.54

Chemical Identifiers

CAS 57771-09-2
Molecular Formula C15H23NO2
Molecular Weight (g/mol) 249.354
InChI Key BWHGDPKOFVNGFQ-UHFFFAOYSA-N
Synonym 4-(Dibutylamino)-2-hydroxybenzaldehyde
PubChem CID 18534534
IUPAC Name 4-(dibutylamino)-2-hydroxybenzaldehyde
SMILES CCCCN(CCCC)C1=CC(=C(C=C1)C=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D32605G
missing translation for 'viewDocuments'
TCI America
D32605G
5 g
Each for $393.32
Only null left
 
D326025G
missing translation for 'viewDocuments'
TCI America
D326025G
25 g
Each for $1,377.54
Only null left
 
Description

Description

Specifications

Chemical Identifiers

57771-09-2
249.354
4-(Dibutylamino)-2-hydroxybenzaldehyde
4-(dibutylamino)-2-hydroxybenzaldehyde
C15H23NO2
BWHGDPKOFVNGFQ-UHFFFAOYSA-N
18534534
CCCCN(CCCC)C1=CC(=C(C=C1)C=O)O

Specifications

57771-09-2
5 g
BWHGDPKOFVNGFQ-UHFFFAOYSA-N
4-(dibutylamino)-2-hydroxybenzaldehyde
18534534
≥96.0% (GC)
4-(Dibutylamino)salicylaldehyde
C15H23NO2
4-(Dibutylamino)-2-hydroxybenzaldehyde
CCCCN(CCCC)C1=CC(=C(C=C1)C=O)O
249.354
249.35
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents