missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dibutylamino)salicylaldehyde 96.0+%, TCI America™
Supplier: TCI America D326025G
Specifications
| 4-(Dibutylamino)salicylaldehyde | |
| C15H23NO2 | |
| 4-(Dibutylamino)-2-hydroxybenzaldehyde | |
| CCCCN(CCCC)C1=CC(=C(C=C1)C=O)O | |
| 249.354 | |
| 249.35 | |
| Liquid |
| 57771-09-2 | |
| 25 g | |
| BWHGDPKOFVNGFQ-UHFFFAOYSA-N | |
| 4-(dibutylamino)-2-hydroxybenzaldehyde | |
| 18534534 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 57771-09-2 | |
| 249.354 | |
| 4-(Dibutylamino)-2-hydroxybenzaldehyde | |
| 4-(dibutylamino)-2-hydroxybenzaldehyde |
| C15H23NO2 | |
| BWHGDPKOFVNGFQ-UHFFFAOYSA-N | |
| 18534534 | |
| CCCCN(CCCC)C1=CC(=C(C=C1)C=O)O |
Safety and Handling
TSCA : No
Recommended Storage : Freezer