missing translation for 'onlineSavingsMsg'
Learn More

4-Deoxypyridoxine hydrochloride, ≥98%, MilliporeSigma™ Supelco™

Catalog No. 111013170
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013170 500 mg
111013169 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013170 Supplier Merck Emd Millipore Supplier No. D0501500MG
Only null left
Vitamins, Nutraceuticals and Natural Products

Specifications

Packaging Poly Bottle
CAS 148-51-6
Physical Form Neat
Linear Formula C8H11 NO2 · HCl
MDL Number MFCD00051296
Melting Point 263°C to 265°C
Type Analytical Standard
Quantity 500 mg
Formula Weight 189.64
Percent Purity ≥98%
Grade Analytical Standard
Show More Show Less