missing translation for 'onlineSavingsMsg'
Learn More

4-Decyloxybenzaldehyde 97.0+%, TCI America™

Catalog No. D11245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11245G 5 g
D112425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11245G Supplier TCI America Supplier No. D11245G

Chemical Identifiers

CAS 24083-16-7
Molecular Formula C17H26O2
Molecular Weight (g/mol) 262.39
MDL Number MFCD00021072
InChI Key WOSYBKJRUQJISL-UHFFFAOYSA-N
PubChem CID 141068
IUPAC Name 4-(decyloxy)benzaldehyde
SMILES CCCCCCCCCCOC1=CC=C(C=O)C=C1

Specifications

Boiling Point 176°C
Quantity 5 g
Synonym 4-decyloxy benzaldehyde, 4-decyloxybenzaldehyde, p-decyloxybenzaldehyde, 4-n-decyloxybenzaldehyde, benzaldehyde,4-decyloxy, benzaldehyde, 4-decyloxy, 4-decylox-ybenzaldehyde, p-n-decyloxybenzaldehyde, acmc-209g8u, p-decyloxy benzaldehyde
Formula Weight 262.39
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Decyloxybenzaldehyde