CAS RN 24083-16-7
IUPAC Name:
4-(decyloxy)benzaldehyde
Synonyms:
4-(decyloxy)benzaldehyde
Molecular Weight (g/mol):
262.39
Molecular Formula:
C17H26O2
InChi Key:
WOSYBKJRUQJISL-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCOC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
4-Decyloxybenzaldehyde 97.0+%, TCI America™
CAS: 24083-16-7 Molecular Formula: C17H26O2 Molecular Weight (g/mol): 262.39 MDL Number: MFCD00021072 InChI Key: WOSYBKJRUQJISL-UHFFFAOYSA-N PubChem CID: 141068 IUPAC Name: 4-(decyloxy)benzaldehyde SMILES: CCCCCCCCCCOC1=CC=C(C=O)C=C1