missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Decanone 97.0+%, TCI America™
$488.61 - $488.61
Chemical Identifiers
| CAS | 624-16-8 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.269 |
| MDL Number | MFCD00009542 |
| InChI Key | MKJDUHZPLQYUCB-UHFFFAOYSA-N |
| Synonym | 4-decanone, hexyl propyl ketone, propyl hexyl ketone, unii-502v1w8kd4, n-propyl n-hexyl ketone, acmc-209n3f, mkjduhzplqyucb-uhfffaoysa, unii-3z25263nt0 component |
| PubChem CID | 12203 |
| IUPAC Name | decan-4-one |
| SMILES | CCCCCCC(=O)CCC |
Chemical Identifiers
| 624-16-8 | |
| 156.269 | |
| MKJDUHZPLQYUCB-UHFFFAOYSA-N | |
| 12203 | |
| CCCCCCC(=O)CCC |
| C10H20O | |
| MFCD00009542 | |
| 4-decanone, hexyl propyl ketone, propyl hexyl ketone, unii-502v1w8kd4, n-propyl n-hexyl ketone, acmc-209n3f, mkjduhzplqyucb-uhfffaoysa, unii-3z25263nt0 component | |
| decan-4-one |
Specifications
| 624-16-8 | |
| 90°C | |
| MFCD00009542 | |
| 4-decanone, hexyl propyl ketone, propyl hexyl ketone, unii-502v1w8kd4, n-propyl n-hexyl ketone, acmc-209n3f, mkjduhzplqyucb-uhfffaoysa, unii-3z25263nt0 component | |
| CCCCCCC(=O)CCC | |
| 156.269 | |
| 156.27 | |
| Liquid |
| Yellow | |
| C10H20O | |
| 25 mL | |
| MKJDUHZPLQYUCB-UHFFFAOYSA-N | |
| decan-4-one | |
| 12203 | |
| ≥97.0% (GC) | |
| 4-Decanone |
Safety and Handling
TSCA : No