missing translation for 'onlineSavingsMsg'
Learn More

4-Decanone 97.0+%, TCI America™

Supplier:  TCI America D153525ML

 View more versions of this product

Catalog No. D153525ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Decanone
Yellow
C10H20O
25 mL
MKJDUHZPLQYUCB-UHFFFAOYSA-N
decan-4-one
12203
≥97.0% (GC)
624-16-8
90°C
MFCD00009542
4-decanone, hexyl propyl ketone, propyl hexyl ketone, unii-502v1w8kd4, n-propyl n-hexyl ketone, acmc-209n3f, mkjduhzplqyucb-uhfffaoysa, unii-3z25263nt0 component
CCCCCCC(=O)CCC
156.269
156.27
Liquid

Chemical Identifiers

624-16-8
156.269
MKJDUHZPLQYUCB-UHFFFAOYSA-N
12203
CCCCCCC(=O)CCC
C10H20O
MFCD00009542
4-decanone, hexyl propyl ketone, propyl hexyl ketone, unii-502v1w8kd4, n-propyl n-hexyl ketone, acmc-209n3f, mkjduhzplqyucb-uhfffaoysa, unii-3z25263nt0 component
decan-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions