missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyclohexylcyclohexanol (cis- and trans- mixture) 98.0+%, TCI America™
$164.03 - $471.60
Chemical Identifiers
| CAS | 2433-14-9 |
|---|---|
| Molecular Formula | C12H22O |
| Molecular Weight (g/mol) | 182.31 |
| MDL Number | MFCD00060071,MFCD13185038 |
| InChI Key | AFKMHDZOVNDWLO-UHFFFAOYSA-N |
| PubChem CID | 75518 |
| IUPAC Name | [1,1'-bi(cyclohexane)]-4-ol |
| SMILES | OC1CCC(CC1)C1CCCCC1 |
Chemical Identifiers
| 2433-14-9 | |
| 182.31 | |
| AFKMHDZOVNDWLO-UHFFFAOYSA-N | |
| [1,1'-bi(cyclohexane)]-4-ol |
| C12H22O | |
| MFCD00060071,MFCD13185038 | |
| 75518 | |
| OC1CCC(CC1)C1CCCCC1 |
Specifications
| 2433-14-9 | |
| C12H22O | |
| 5 g | |
| OC1CCC(CC1)C1CCCCC1 | |
| 182.31 | |
| 182.31 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00060071,MFCD13185038 | |
| AFKMHDZOVNDWLO-UHFFFAOYSA-N | |
| [1,1'-bi(cyclohexane)]-4-ol | |
| 75518 | |
| ≥98.0% (GC) | |
| 4-Cyclohexylcyclohexanol (cis- and trans- mixture) |
Safety and Handling
TSCA : No