accessibility menu, dialog, popup

UserName

CAS RN 2433-14-9

OH

CAS RN 2433-14-9

IUPAC Name: [1,1'-bi(cyclohexane)]-4-ol
Synonyms: [1,1'-bi(cyclohexane)]-4-ol
Molecular Weight (g/mol): 182.31
Molecular Formula: C12H22O
InChi Key: AFKMHDZOVNDWLO-UHFFFAOYSA-N
SMILES: OC1CCC(CC1)C1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results