missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylbenzoic Acid 98.0+%, TCI America™

Catalog No. p-7125037
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
C15235G 5 g
C152325G 25 g
2 options

Catalog No. C15235G

Supplier: TCI America C15235G

Only null left

Chemical Identifiers

CAS 20029-52-1
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00039459
InChI Key QCIWHVKGVVQHIY-UHFFFAOYSA-N
Synonym 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane
PubChem CID 88337
IUPAC Name 4-cyclohexylbenzoic acid
SMILES C1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 200°C
Color White
Quantity 5 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyclohexylbenzoic Acid