missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylbenzoic Acid 98.0+%, TCI America™

Catalog No. C15235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15235G 5 g
C152325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15235G Supplier TCI America Supplier No. C15235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20029-52-1
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00039459
InChI Key QCIWHVKGVVQHIY-UHFFFAOYSA-N
Synonym 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane
PubChem CID 88337
IUPAC Name 4-cyclohexylbenzoic acid
SMILES C1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 200°C
Color White
Quantity 5 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyclohexylbenzoic Acid