Learn More
4-Cyclohexylbenzoic acid, 98%
CAS: 20029-52-1 | C13H16O2 | 204.269 g/mol
$89.32 - $909.14
Chemical Identifiers
| CAS | 20029-52-1 |
|---|---|
| Molecular Formula | C13H16O2 |
| Molecular Weight (g/mol) | 204.269 |
| MDL Number | MFCD00039459 |
| InChI Key | QCIWHVKGVVQHIY-UHFFFAOYSA-N |
| Synonym | 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane |
| PubChem CID | 88337 |
| IUPAC Name | 4-cyclohexylbenzoic acid |
| SMILES | C1CCC(CC1)C2=CC=C(C=C2)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0508604
|
Thermo Scientific Chemicals
L0508604 |
2 g |
Each for $89.32
|
|
|||||
|
AAL0508609
|
Thermo Scientific Chemicals
L0508609 |
10 g |
Each for $247.14
|
|
|||||
|
AAL0508618
|
Thermo Scientific Chemicals
L0508618 |
50 g |
Each for $909.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 20029-52-1 | |
| 204.269 | |
| QCIWHVKGVVQHIY-UHFFFAOYSA-N | |
| 88337 | |
| C1CCC(CC1)C2=CC=C(C=C2)C(=O)O |
| C13H16O2 | |
| MFCD00039459 | |
| 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane | |
| 4-cyclohexylbenzoic acid |
Specifications
| 20029-52-1 | |
| C13H16O2 | |
| 2 g | |
| 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane | |
| C1CCC(CC1)C2=CC=C(C=C2)C(=O)O | |
| 204.269 | |
| 204.27 | |
| 4-Cyclohexylbenzoic acid |
| 194°C to 198°C | |
| MFCD00039459 | |
| 1966671 | |
| QCIWHVKGVVQHIY-UHFFFAOYSA-N | |
| 4-cyclohexylbenzoic acid | |
| 88337 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 243-472-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only