missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylbenzoic acid, 98%

Catalog No. AAL0508618
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0508618 50 g
AAL0508604 2 g
AAL0508609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0508618 Supplier Thermo Scientific Chemicals Supplier No. L0508618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20029-52-1 | C13H16O2 | 204.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20029-52-1
Molecular Formula C13H16O2
Molecular Weight (g/mol) 204.269
MDL Number MFCD00039459
InChI Key QCIWHVKGVVQHIY-UHFFFAOYSA-N
Synonym 4-cyclohexyl-benzoic acid, benzoic acid, 4-cyclohexyl, 4cyclohexylbenzoic acid, 4-cyclohexylbenzoicacid, p-cyclohexylbenzoic acid, acmc-1cmbu, cyclohexybenzoicacid, 4-carboxyphenyl cyclohexane
PubChem CID 88337
IUPAC Name 4-cyclohexylbenzoic acid
SMILES C1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 194°C to 198°C
Quantity 50 g
Beilstein 1966671
Formula Weight 204.27
Percent Purity 98%
Chemical Name or Material 4-Cyclohexylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-472-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only