missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylbenzenesulfonyl chloride, 97%

Catalog No. AAH3361609
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3361609 10 g
AAH3361603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3361609 Supplier Thermo Scientific Chemicals Supplier No. H3361609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56354-57-5 | C12H15ClO2S | 258.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56354-57-5
Molecular Formula C12H15ClO2S
Molecular Weight (g/mol) 258.76
MDL Number MFCD00625713
InChI Key CREMYEDHKUWVTB-UHFFFAOYSA-N
Synonym 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
PubChem CID 4339442
IUPAC Name 4-cyclohexylbenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1

Specifications

Melting Point 52°C to 54°C
Quantity 10 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 258.77
Percent Purity 97%
Chemical Name or Material 4-Cyclohexylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only