accessibility menu, dialog, popup

UserName

CAS RN 56354-57-5

IUPAC Name: 4-cyclohexylbenzene-1-sulfonyl chloride
Synonyms: 4-cyclohexylbenzenesulfonyl chloride
Molecular Weight (g/mol): 258.76
Molecular Formula: C12H15ClO2S
InChi Key: CREMYEDHKUWVTB-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)