missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyclohexanebutyric Acid 98.0+%, TCI America™
Supplier: TCI America C095225G
Specifications
| 4-Cyclohexanebutyric Acid | |
| White-Yellow | |
| MFCD00001529 | |
| 4-cyclohexylbutyric acid, cyclohexanebutyric acid, cyclohexanebutanoic acid, 4-cyclohexanebutyric acid, cyclohexanebutyricacid, pubchem15521, acmc-20aj2k, cyclohexane-butyric acid, 4-cyclohexanebutyricacid, 4-cyclohexyl butyric acid | |
| C1CCC(CC1)CCCC(=O)O | |
| 170.252 | |
| 170.25 | |
| Crystalline Lumps |
| 4441-63-8 | |
| C10H18O2 | |
| 25 g | |
| UVZMNGNFERVGRC-UHFFFAOYSA-N | |
| 4-cyclohexylbutanoic acid | |
| 75200 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 4441-63-8 | |
| 170.252 | |
| UVZMNGNFERVGRC-UHFFFAOYSA-N | |
| 75200 | |
| C1CCC(CC1)CCCC(=O)O |
| C10H18O2 | |
| MFCD00001529 | |
| 4-cyclohexylbutyric acid, cyclohexanebutyric acid, cyclohexanebutanoic acid, 4-cyclohexanebutyric acid, cyclohexanebutyricacid, pubchem15521, acmc-20aj2k, cyclohexane-butyric acid, 4-cyclohexanebutyricacid, 4-cyclohexyl butyric acid | |
| 4-cyclohexylbutanoic acid |
Safety and Handling
TSCA : Yes