accessibility menu, dialog, popup

UserName

HOO

CAS RN 4441-63-8

IUPAC Name: 4-cyclohexylbutanoic acid
Synonyms: 4-cyclohexylbutanoic acid
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: UVZMNGNFERVGRC-UHFFFAOYSA-N
SMILES: OC(=O)CCCC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results