missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexanebutyric Acid 98.0+%, TCI America™

Catalog No. C09525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09525G 5 g
C095225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09525G Supplier TCI America Supplier No. C09525G

Chemical Identifiers

CAS 4441-63-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00001529
InChI Key UVZMNGNFERVGRC-UHFFFAOYSA-N
Synonym 4-cyclohexylbutyric acid, cyclohexanebutyric acid, cyclohexanebutanoic acid, 4-cyclohexanebutyric acid, cyclohexanebutyricacid, pubchem15521, acmc-20aj2k, cyclohexane-butyric acid, 4-cyclohexanebutyricacid, 4-cyclohexyl butyric acid
PubChem CID 75200
IUPAC Name 4-cyclohexylbutanoic acid
SMILES C1CCC(CC1)CCCC(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Cyclohexanebutyric Acid