missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanopyridine 98.0+%, TCI America™
Supplier: TCI America C0457100G
Specifications
| 4-Cyanopyridine | |
| 80°C | |
| 198°C | |
| MFCD00006417 | |
| 4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155 | |
| C1=CN=CC=C1C#N | |
| 104.112 | |
| CHEBI:28020 | |
| ≥98.0% (GC,T) |
| 100-48-1 | |
| White | |
| C6H4N2 | |
| 100 g | |
| GPHQHTOMRSGBNZ-UHFFFAOYSA-N | |
| pyridine-4-carbonitrile | |
| 7506 | |
| 104.11 | |
| Crystalline Powder |
Chemical Identifiers
| 100-48-1 | |
| 104.112 | |
| GPHQHTOMRSGBNZ-UHFFFAOYSA-N | |
| 7506 | |
| pyridine-4-carbonitrile |
| C6H4N2 | |
| MFCD00006417 | |
| 4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155 | |
| CHEBI:28020 | |
| C1=CN=CC=C1C#N |
Safety and Handling
EINECSNumber : (5)-0743
TSCA : Yes