missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopyridine 98.0+%, TCI America™

Catalog No. p-7124434
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C045725G 25 g
C0457100G 100 g
C0457500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C045725G Supplier TCI America Supplier No. C045725G
Only null left

Chemical Identifiers

CAS 100-48-1
Molecular Formula C6H4N2
Molecular Weight (g/mol) 104.112
MDL Number MFCD00006417
InChI Key GPHQHTOMRSGBNZ-UHFFFAOYSA-N
Synonym 4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
PubChem CID 7506
ChEBI CHEBI:28020
IUPAC Name pyridine-4-carbonitrile
SMILES C1=CN=CC=C1C#N

Specifications

Melting Point 80°C
Color White
Boiling Point 198°C
Quantity 25 g
Formula Weight 104.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanopyridine
EINECSNumber (5)-0743
TSCA Yes