missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopiperidine, 96%

Catalog No. AAH2772206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2772206 5 g
AAH2772214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2772206 Supplier Thermo Scientific Chemicals Supplier No. H2772206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4395-98-6 | C6H10N2 | 110.16 g/mol

4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4395-98-6
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD05022468
InChI Key FSDNTQSJGHSJBG-UHFFFAOYSA-N
Synonym 4-cyanopiperidine, 4-piperidinecarbonitrile, 4-cyanopiperdine, 4-cyanopyperidine, 4-cyano piperdine, 4-cyano piperidine, 4-cyano-piperidine, acmc-20aipl, pubchem9349
PubChem CID 138223
IUPAC Name piperidine-4-carbonitrile
SMILES C1CNCCC1C#N

Specifications

Density 0.98
Boiling Point 226°C
Flash Point 91°C (196°F)
Odor Amine-like
Refractive Index 1.4755
Quantity 5 g
UN Number UN3276
Solubility Information Soluble in chloroform.
Formula Weight 110.16
Percent Purity 96%
Chemical Name or Material 4-Cyanopiperidine
Show More Show Less
Hazard Category H302-H311+H331-H315-H318
Hazard Statement GHS H Statement
H311-H227-H302-H332-H315-H319-H335
Toxic in contact with skin.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only