missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$113.59 - $334.98
Chemical Identifiers
| CAS | 126747-14-6 |
|---|---|
| Molecular Formula | C7H6BNO2 |
| Molecular Weight (g/mol) | 146.94 |
| MDL Number | MFCD01318968 |
| InChI Key | CEBAHYWORUOILU-UHFFFAOYSA-N |
| Synonym | 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile |
| PubChem CID | 2734326 |
| IUPAC Name | (4-cyanophenyl)boronic acid |
| SMILES | OB(O)C1=CC=C(C=C1)C#N |
Chemical Identifiers
| 126747-14-6 | |
| 146.94 | |
| CEBAHYWORUOILU-UHFFFAOYSA-N | |
| 2734326 | |
| OB(O)C1=CC=C(C=C1)C#N |
| C7H6BNO2 | |
| MFCD01318968 | |
| 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile | |
| (4-cyanophenyl)boronic acid |
Specifications
| 126747-14-6 | |
| C7H6BNO2 | |
| 5 g | |
| 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile | |
| OB(O)C1=CC=C(C=C1)C#N | |
| 146.94 | |
| 146.94 |
| White-Yellow | |
| MFCD01318968 | |
| 2811 | |
| CEBAHYWORUOILU-UHFFFAOYSA-N | |
| (4-cyanophenyl)boronic acid | |
| 2734326 | |
| Crystalline Powder |
Safety and Handling
TSCA : No