missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenylboronic acid, 97%

CAS: 126747-14-6 | C7H6BNO2 | 146.94 g/mol

$111.90 - $782.59

Chemical Identifiers

CAS 126747-14-6
Molecular Formula C7H6BNO2
Molecular Weight (g/mol) 146.94
MDL Number MFCD01318968
InChI Key CEBAHYWORUOILU-UHFFFAOYSA-N
Synonym 4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid
PubChem CID 2734326
IUPAC Name (4-cyanophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC359040010
SDS View Documents
Thermo Scientific Chemicals
359040010
1 g Glass bottle
Each for $111.90
Only null left
 
AC359040100
SDS View Documents
Thermo Scientific Chemicals
359040100
10 g Glass bottle
Each for $782.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

126747-14-6
146.94
CEBAHYWORUOILU-UHFFFAOYSA-N
2734326
OB(O)C1=CC=C(C=C1)C#N
C7H6BNO2
MFCD01318968
4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid
(4-cyanophenyl)boronic acid

Specifications

126747-14-6
100.0
White to Yellow
96% min. (HPLC)
C7H6BNO2
1 g
Solubility in water: insoluble
OB(O)C1=CC=C(C=C1)C#N
146.94
146.94
Powder
96.0
>300°C
Authentic
Glass bottle
MFCD01318968
4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid
CEBAHYWORUOILU-UHFFFAOYSA-N
(4-cyanophenyl)boronic acid
2734326
97%
4-Cyanophenylboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only