missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenyl isothiocyanate, 98%

Catalog No. p-7045297
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1017303 1 g
AAL1017306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1017303 Supplier Thermo Scientific Chemicals Supplier No. L1017303
Only null left

CAS: 2719-32-6 | C8H4N2S | 160.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2719-32-6
Molecular Formula C8H4N2S
Molecular Weight (g/mol) 160.194
MDL Number MFCD00041085
InChI Key DZFKAXLNKZXNHD-UHFFFAOYSA-N
Synonym 4-cyanophenyl isothiocyanate, benzonitrile, 4-isothiocyanato, p-cyanophenyl isothiocyanate, 4-cyanophenylisothiocyanate, 4-cyano-phenyl-isothiocyanate, 4-isothiocyanatobenzenenitrile, 4-isothiocyanato-benzonitrile, benzonitrile,4-isothiocyanato, isothiocyanic acid, p-cyanophenyl ester, benzenenitrile, 4-isothiocyanato
PubChem CID 17626
IUPAC Name 4-isothiocyanatobenzonitrile
SMILES C1=CC(=CC=C1C#N)N=C=S

Specifications

Melting Point 118°C to 122°C
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2802621
Sensitivity Moisture sensitive
Formula Weight 160.2
Percent Purity 98%
Chemical Name or Material 4-Cyanophenyl isothiocyanate
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 220-323-2
RTECSNumber NX8479500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only