missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenyl isocyanate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 405970250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC405970250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

40465-45-0
144.13
TVSXDZNUTPLDKY-UHFFFAOYSA-N
170380
C1=CC(=CC=C1C#N)N=C=O
C8H4N2O
MFCD00037038
4-cyanophenyl isocyanate, benzonitrile, 4-isocyanato, 4-cyanophenylisocyanate, p-cyanophenyl isocyanate, benzonitrile,4-isocyanato, 4-isocyanatobenzenecarbonitrile, 4-cyanophenylisocyanat, p-cyanophenylisocyanate, #, 1-cyano-4-isocyanato-benzene
4-isocyanatobenzonitrile

Specifications

4-Cyanophenyl isocyanate
94.0
Authentic
Glass bottle
NCC6H4NCO
MFCD00037038
Solubility in water: insoluble (dec). Other solubilities: soluble in toluene and chloroform
C1=CC(=CC=C1C#N)N=C=O
144.13
144.13
40465-45-0
100.0
94% min. (GC)
C8H4N2O
25g
4-cyanophenyl isocyanate, benzonitrile, 4-isocyanato, 4-cyanophenylisocyanate, p-cyanophenyl isocyanate, benzonitrile,4-isocyanato, 4-isocyanatobenzenecarbonitrile, 4-cyanophenylisocyanat, p-cyanophenylisocyanate, #, 1-cyano-4-isocyanato-benzene
TVSXDZNUTPLDKY-UHFFFAOYSA-N
4-isocyanatobenzonitrile
170380
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

EINECSNumber : 254-934-

SDS
Documents

Documents

Promotions

Promotions