missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenyl isocyanate, 95%, Thermo Scientific Chemicals

Catalog No. p-3778065
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405970050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC405970050 Supplier Thermo Scientific Chemicals Supplier No. 405970050

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40465-45-0
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.13
MDL Number MFCD00037038
InChI Key TVSXDZNUTPLDKY-UHFFFAOYSA-N
PubChem CID 170380
IUPAC Name 4-isocyanatobenzonitrile
SMILES C1=CC(=CC=C1C#N)N=C=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 102°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula NCC6H4NCO
Quantity 5 g
Synonym 4-cyanophenyl isocyanate, benzonitrile, 4-isocyanato, 4-cyanophenylisocyanate, p-cyanophenyl isocyanate, benzonitrile,4-isocyanato, 4-isocyanatobenzenecarbonitrile, 4-cyanophenylisocyanat, p-cyanophenylisocyanate, #, 1-cyano-4-isocyanato-benzene
Solubility Information Solubility in water: insoluble (dec). Other solubilities: soluble in toluene and chloroform
Formula Weight 144.13
Percent Purity 95%
Physical Form Crystalline Needles or Solid
Chemical Name or Material 4-Cyanophenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 254-934-

RUO – Research Use Only