missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenyl 4-(3-Butenyloxy)benzoate 97.0+%, TCI America™

Catalog No. B15861G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15861G Supplier TCI America Supplier No. B15861G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 114482-57-4
Molecular Formula C18H15NO3
Molecular Weight (g/mol) 293.322
MDL Number MFCD00191384
InChI Key XGRKLNHEYFRXFW-UHFFFAOYSA-N
Synonym 4-(3-Butenyloxy)benzoic Acid 4-Cyanophenyl Ester
PubChem CID 600247
IUPAC Name (4-cyanophenyl) 4-but-3-enoxybenzoate
SMILES C=CCCOC1=CC=C(C=C1)C(=O)OC2=CC=C(C=C2)C#N

Specifications

Melting Point 84°C
Quantity 1 g
Formula Weight 293.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanophenyl 4-(3-Butenyloxy)benzoate