missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanophenacyl Bromide 96.0+%, TCI America™
Supplier: TCI America C134825G
Specifications
| 4-Cyanophenacyl Bromide | |
| 93°C | |
| C9H6BrNO | |
| 25 g | |
| 4-2-bromoacetyl benzonitrile, 4-cyanophenacyl bromide, 4-bromoacetyl benzonitrile, 2-bromo-4'-cyanoacetophenone, benzonitrile, 4-bromoacetyl, p-cyanophenacyl bromide, 4-2-bromo-acetyl-benzonitrile, 4-2-bromoethanoyl benzonitrile, 4-2-bromoacetyl benzenecarbonitrile | |
| C1=CC(=CC=C1C#N)C(=O)CBr | |
| 224.057 | |
| 224.06 | |
| Crystalline Powder |
| 20099-89-2 | |
| White-Yellow | |
| MFCD00052931 | |
| 2923 | |
| LJANCPRIUMHGJE-UHFFFAOYSA-N | |
| 4-(2-bromoacetyl)benzonitrile | |
| 98654 | |
| ≥96.0% (GC,T) |
Chemical Identifiers
| 20099-89-2 | |
| 224.057 | |
| LJANCPRIUMHGJE-UHFFFAOYSA-N | |
| 98654 | |
| C1=CC(=CC=C1C#N)C(=O)CBr |
| C9H6BrNO | |
| MFCD00052931 | |
| 4-2-bromoacetyl benzonitrile, 4-cyanophenacyl bromide, 4-bromoacetyl benzonitrile, 2-bromo-4'-cyanoacetophenone, benzonitrile, 4-bromoacetyl, p-cyanophenacyl bromide, 4-2-bromo-acetyl-benzonitrile, 4-2-bromoethanoyl benzonitrile, 4-2-bromoacetyl benzenecarbonitrile | |
| 4-(2-bromoacetyl)benzonitrile |
Safety and Handling
TSCA : No