accessibility menu, dialog, popup

UserName

BrON

CAS RN 20099-89-2

IUPAC Name: 4-(2-bromoacetyl)benzonitrile
Synonyms: 4-(2-bromoacetyl)benzonitrile
Molecular Weight (g/mol): 224.06
Molecular Formula: C9H6BrNO
InChi Key: LJANCPRIUMHGJE-UHFFFAOYSA-N
SMILES: BrCC(=O)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)