missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyanomethyl)benzeneboronic acid, 95%

Catalog No. AAL1995506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1995506 5 g
AAL1995503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1995506 Supplier Thermo Scientific Chemicals Supplier No. L1995506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91983-26-5 | C8H8BNO2 | 160.967 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91983-26-5
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.967
MDL Number MFCD01632200
InChI Key YKVMTTIYBSVPEQ-UHFFFAOYSA-N
Synonym 4-cyanomethyl benzeneboronic acid, 4-cyanomethyl phenyl boronic acid, 4-cyanomethylphenylboronic acid, 4-cyanomethylphenyl boronic acid, 4-cyanomethyl phenylboronic acid, 4-boronophenylacetonitrile, boronic acid, 4-cyanomethyl phenyl, pubchem7789, acmc-209ree
PubChem CID 2773342
IUPAC Name [4-(cyanomethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CC#N)(O)O

Specifications

Melting Point ∼196°C to 200°C
Quantity 5 g
Beilstein 3031323
Formula Weight 160.97
Percent Purity 98%
Chemical Name or Material 4-(Cyanomethyl)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only