missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanoindole 98.0+%, TCI America™

Catalog No. C20831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20831G 1 g
C20835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20831G Supplier TCI America Supplier No. C20831G

Chemical Identifiers

CAS 16136-52-0
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00152045
InChI Key CEUFGDDOMXCXFW-UHFFFAOYSA-N
Synonym 4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide
PubChem CID 3817602
IUPAC Name 1H-indole-4-carbonitrile
SMILES C1=CC(=C2C=CNC2=C1)C#N

Specifications

Melting Point 120°C
Quantity 1 g
UN Number 3439
Formula Weight 142.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanoindole