accessibility menu, dialog, popup

UserName

NHN

CAS RN 16136-52-0

IUPAC Name: 1H-indole-4-carbonitrile
Synonyms: indole-4-cyano-
Molecular Weight (g/mol): 142.16
Molecular Formula: C9H6N2
InChi Key: CEUFGDDOMXCXFW-UHFFFAOYSA-N
SMILES: N#CC1=C2C=CNC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)