missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanoindole, 99%

Catalog No. AAH6339103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6339103 1 g
AAH6339106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6339103 Supplier Thermo Scientific Chemicals Supplier No. H6339103

CAS: 16136-52-0 | C9H6N2 | 142.161 g/mol

4-Cyanoindole is used as a synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanoindole is used as a synthetic intermediate.

Solubility
Soluble in water.

Notes
Heat, light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids.

Chemical Identifiers

CAS 16136-52-0
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00152045
InChI Key CEUFGDDOMXCXFW-UHFFFAOYSA-N
Synonym 4-cyanoindole, 4-indolecarbonitrile, indole-4-carbonitrile, indole-4-cyano, 4-cyano-1h-indole, 4-cyano indole, pubchem7327, acmc-209dmi, 1h-indol-4-yl cyanide
PubChem CID 3817602
IUPAC Name 1H-indole-4-carbonitrile
SMILES C1=CC(=C2C=CNC2=C1)C#N

Specifications

Melting Point 120°C to 125°C
Color Gray
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 142.16
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 4-Cyanoindole
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H318-H302-H317-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only