missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyanobenzylidene-4-ethoxyaniline 98.0+%, TCI America™

Catalog No. C07441G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C07441G Supplier TCI America Supplier No. C07441G
Only null left

Chemical Identifiers

CAS 34128-02-4
Molecular Formula C16H14N2O
Molecular Weight (g/mol) 250.301
MDL Number MFCD00059585
InChI Key WTTZYFPFXIZAOS-UHFFFAOYSA-N
PubChem CID 613590
IUPAC Name 4-[(4-ethoxyphenyl)iminomethyl]benzonitrile
SMILES CCOC1=CC=C(C=C1)N=CC2=CC=C(C=C2)C#N

Specifications

Quantity 1 g
Formula Weight 250.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Cyanobenzylidene-4-ethoxyaniline