missing translation for 'onlineSavingsMsg'
Learn More

(4-Cyanobenzyl)triphenylphosphonium chloride, 99%

Catalog No. p-7045840
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1218706 5 g
AAL1218714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1218706 Supplier Thermo Scientific Chemicals Supplier No. L1218706
Only null left

CAS: 20430-33-5 | C26H21ClNP | 413.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20430-33-5
Molecular Formula C26H21ClNP
Molecular Weight (g/mol) 413.88
MDL Number MFCD00011752
InChI Key GXMFPJIKPZDNCS-UHFFFAOYSA-M
Synonym 4-cyanobenzyl triphenylphosphonium chloride, 4-cyanophenyl methyl triphenylphosphanium chloride, 4-triphenylphosphino methyl benzenecarbonitrile, chloride, acmc-20apna, phosphonium, 4-cyanophenyl methyl triphenyl-, chloride, 4-cyanobenzyltriphenylphosphonium chloride, p-cyanobenzyl-triphenylphosphonium chloride, 4-cyanobenzyl triphenylphosphoniumchloride, 4-cyanobenzyl-triphenylphosphonium chloride
PubChem CID 2724468
SMILES [Cl-].N#CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 322°C to 326°C
Odor Odorless
Quantity 5 g
Beilstein 4630137
IUPAC Name (4-cyanophenyl)methyl-triphenylphosphanium;chloride
Formula Weight 413.89
Percent Purity 99%
Chemical Name or Material (4-Cyanobenzyl)triphenylphosphonium chloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only