missing translation for 'onlineSavingsMsg'
Learn More

(4-Cyanobenzyl)triphenylphosphonium chloride, 99%

Catalog No. aal1218714
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL1218714 25 g
AAL1218706 5 g
2 options

Catalog No. AAL1218714

Supplier: Thermo Scientific Chemicals L1218714

Only null left

CAS: 20430-33-5 | C26H21ClNP | 413.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20430-33-5
Molecular Formula C26H21ClNP
Molecular Weight (g/mol) 413.88
MDL Number MFCD00011752
InChI Key GXMFPJIKPZDNCS-UHFFFAOYSA-M
Synonym 4-cyanobenzyl triphenylphosphonium chloride, 4-cyanophenyl methyl triphenylphosphanium chloride, 4-triphenylphosphino methyl benzenecarbonitrile, chloride, acmc-20apna, phosphonium, 4-cyanophenyl methyl triphenyl-, chloride, 4-cyanobenzyltriphenylphosphonium chloride, p-cyanobenzyl-triphenylphosphonium chloride, 4-cyanobenzyl triphenylphosphoniumchloride, 4-cyanobenzyl-triphenylphosphonium chloride
PubChem CID 2724468
SMILES [Cl-].N#CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 322°C to 326°C
Odor Odorless
Quantity 25 g
Beilstein 4630137
IUPAC Name [(4-cyanophenyl)methyl]triphenylphosphanium chloride
Formula Weight 413.89
Percent Purity 99%
Chemical Name or Material (4-Cyanobenzyl)triphenylphosphonium chloride
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only