missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7124953
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C139825G 25 g
C1398250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C139825G Supplier TCI America Supplier No. C139825G

Chemical Identifiers

CAS 17201-43-3
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00001829
InChI Key UMLFTCYAQPPZER-UHFFFAOYSA-N
Synonym 4-cyanobenzyl bromide, 4-bromomethyl benzonitrile, p-cyanobenzyl bromide, alpha-bromo-p-tolunitrile, benzonitrile, 4-bromomethyl, 4-bromomethyl-benzonitrile, alpha-bromo-p-toluonitrile, p-bromomethyl benzonitrile, 4-cyano benzyl bromide, benzonitrile, p-bromomethyl
PubChem CID 86996
IUPAC Name 4-(bromomethyl)benzonitrile
SMILES C1=CC(=CC=C1CBr)C#N

Specifications

Melting Point 116°C
Color White
Boiling Point 143°C
Quantity 25 g
UN Number 3261
Formula Weight 196.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzyl Bromide