Learn More
4-Cyanobenzophenone, 98%
CAS: 1503-49-7 | C14H9NO | 207.232 g/mol
Supplier: Thermo Scientific Chemicals A1576406
Description
Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)quadricyclane(Q) and QN isomerization. It is employed as a triplet sensitizers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSubstituted benzophenones are used to determine the quantum yields for norbornadiene(N)→quadricyclane(Q) and Q→N isomerization. It is employed as a triplet sensitizers.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
| 4-Cyanobenzophenone | |
| 110°C to 114°C | |
| MFCD00016382 | |
| UN3439 | |
| 4-cyanobenzophenone, 4-benzoyl-benzonitrile, benzonitrile, 4-benzoyl, 4-cyano-benzophenon, 4-cyano-benzophenone, acmc-20amk8, #, benzonitrile,4-benzoyl, 4-phenylcarbonyl benzenecarbonitrile | |
| YSZWJJANSNFQMM-UHFFFAOYSA-N | |
| 4-benzoylbenzonitrile | |
| 73921 | |
| 98% |
| 1503-49-7 | |
| C14H9NO | |
| 5 g | |
| 641181 | |
| Insoluble in water. | |
| C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N | |
| 207.232 | |
| 207.23 |
Chemical Identifiers
| 1503-49-7 | |
| 207.232 | |
| YSZWJJANSNFQMM-UHFFFAOYSA-N | |
| 4-benzoylbenzonitrile |
| C14H9NO | |
| MFCD00016382 | |
| 73921 | |
| C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 216-126-6
TSCA : No
RUO – Research Use Only