missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzenesulfonamide, 97%

Catalog No. AAH3183603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3183603 1 g
AAH3183606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3183603 Supplier Thermo Scientific Chemicals Supplier No. H3183603

CAS: 3119-02-6 | C7H6N2O2S | 182.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3119-02-6
MDL Number MFCD02089633

Specifications

Melting Point 168°C to 171°C
Odor Odorless
Molecular Formula C7H6N2O2S
Quantity 1 g
Synonym 4-cyanobenzene-1-sulfonamide, p-cyanobenzenesulfonamide, 4-cyanobenzenesulphonamide, benzenesulfonamide, p-cyano, 4-cyano-benzenesulfonamide, benzenesulfonamide, 4-cyano, p-cyanobenzenesulphonamide, benzenesulfonamide, 4-cyano-9ci, 4-sulphamoylbenzonitrile, wln: zswr dcn
InChI Key UZECCNDOASGYNH-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C#N)S(=O)(=O)N
IUPAC Name 4-cyanobenzenesulfonamide
Molecular Weight (g/mol) 182.20
PubChem CID 76566
Formula Weight 182.2
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzenesulfonamide
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-492-5
RTECSNumber DB1582000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only