accessibility menu, dialog, popup

UserName

CAS RN 3119-02-6

IUPAC Name: 4-cyanobenzene-1-sulfonamide
Synonyms: P-cyanobenzenesulfonamide
Molecular Weight (g/mol): 182.2
Molecular Formula: C7H6N2O2S
InChi Key: UZECCNDOASGYNH-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)