missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzeneboronic acid, 98%

CAS: 126747-14-6 | C7H6BNO2 | 146.94 g/mol

Supplier:  Thermo Scientific Chemicals L1800703

 View more versions of this product

Catalog No. AAL1800703


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Cyanobenzeneboronic acid
>300°C
MFCD01318968
6593772
CEBAHYWORUOILU-UHFFFAOYSA-N
(4-cyanophenyl)boronic acid
2734326
98%
126747-14-6
C7H6BNO2
1 g
4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid
OB(O)C1=CC=C(C=C1)C#N
146.94
146.94

Chemical Identifiers

126747-14-6
146.94
CEBAHYWORUOILU-UHFFFAOYSA-N
2734326
OB(O)C1=CC=C(C=C1)C#N
C7H6BNO2
MFCD01318968
4-cyanophenyl boronic acid, 4-cyanobenzeneboronic acid, 4-dihydroxyboranyl benzonitrile, 4-cyano-phenyl-boronic acid, 4-boronobenzonitrile, p-cyanophenylboronic acid, 4-cyanobenzene boronic acid, boronic acid, 4-cyanophenyl, 4-cyano-phenyl boronic acid
(4-cyanophenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only