missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzaldehyde, 98+%

Catalog No. p-7022235
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1491406 5 g
AAA1491414 25 g
AAA1491422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1491406 Supplier Thermo Scientific Chemicals Supplier No. A1491406
Only null left

CAS: 105-07-7 | C8H5NO | 131.13 g/mol

4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanobenzaldehyde, is used as an intermediate for organic synthesis.It is also used as an intermediate in the pharmaceutical and research department.

Solubility
It is soluble in water, ethanol, ether and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong acids, Strong bases, Strong reducing agents.

Chemical Identifiers

CAS 105-07-7
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.13
MDL Number MFCD00003376
InChI Key WZWIQYMTQZCSKI-UHFFFAOYSA-N
Synonym 4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile
PubChem CID 66042
IUPAC Name 4-formylbenzonitrile
SMILES O=CC1=CC=C(C=C1)C#N

Specifications

Melting Point 99°C to 103°C
Boiling Point 133°C (12 mmHg)
Odor Characteristic
Quantity 5 g
Beilstein 606473
Sensitivity Air Sensitive
Solubility Information It is soluble in water,ethanol,ether and chloroform.
Formula Weight 131.13
Percent Purity ≥98%
Chemical Name or Material 4-Cyanobenzaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-267-3
RTECSNumber CU5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only