missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzaldehyde, 98%

Catalog No. AC110720050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC110720050 5 g Glass bottle
AC110720250 25 g Glass bottle
AC110721000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC110720050 Supplier Thermo Scientific Chemicals Supplier No. 110720050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 105-07-7 | C8H5NO | 131.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105-07-7
Molecular Formula C8H5NO
Molecular Weight (g/mol) 131.13
MDL Number MFCD00003376
InChI Key WZWIQYMTQZCSKI-UHFFFAOYSA-N
Synonym 4-cyanobenzaldehyde, benzonitrile, 4-formyl, p-cyanobenzaldehyde, p-formylbenzonitrile, benzaldehyde, p-cyano, benzonitrile, p-formyl, usaf kf-1, terephthalaldehydonitrile, p-cyanobenzenecarboxaldehyde, 4-formyl-benzonitrile
PubChem CID 66042
IUPAC Name 4-formylbenzonitrile
SMILES O=CC1=CC=C(C=C1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 100°C to 101°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NCC6H4CHO
Quantity 5 g
Beilstein 10, 672
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,methylene chloride and,ethylacetate
Formula Weight 131.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanobenzaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-267-3
RTECSNumber CU5250000
TSCA TSCA

RUO – Research Use Only